Tubing connectors, straight
Supplier: KARTELL
Certificates
About this item
PP, white.
- Good mechanical strength and chemical resistance
- Very good thermal resistance
- Autoclavable
- Conforms to Food and Drug regulations 90/128 CEE, 92/39 CEE and FDA 21 CFR 177.1520
With tapered ends, for connecting tubing with the same diameter.