ContactUsLinkComponent
Condensers, double surface, with Quickfit® ground joints
Condensers, double surface, with Quickfit® ground joints
Catalog # BIBBCX5/25/SC
Supplier:  SciLabware
undefined
Condensers, double surface, with Quickfit® ground joints
Catalog # BIBBCX5/25/SC
Supplier:  SciLabware
Specifications
  • Cone size:
    34/35 NS
  • Jacket length:
    205 mm
  • Pk:
    2
Specifications
About this item
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
With ground cone. Highly efficient condensers with large cooling surface. Inner cooling surface gives “baffle” effect. Outer cooling surface prevents “creep” with low boiling point liquids. Detachable plastic screwthread connectors provide easy and safe attachment of flexible tubing.