Condensers, Allihn, with Quickfit® ground joints
Supplier: SciLabware
Certificates
About this item
Condensers acc. to Allihn, PYREX® borosilicate glass, for condensing vapours.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for reflux applications. With NS cone. Inner bulb sections provide increased cooling surface. With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing.
Frequently Bought Together