Orders LinkContactUsLinkComponent
Cation exchange HPLC columns, PRP-X200
Cation exchange HPLC columns, PRP-X200
Supplier:   HAMILTON BONADUZ
Cation exchange HPLC columns, PRP-X200
Supplier:  HAMILTON BONADUZ
Certificates
Cation exchange HPLC columns, PRP-X200
Cation exchange HPLC columns, PRP-X200
14 Options Available Below
Product Details & Documents
Polymeric cation exchange packing for separation of inorganic and organic cations.
  • pH stable from 1 to 13
  • Easily seperates mono or divalent cations, alcalins et alcalino-terreux
  • Use with organic solvent from 0 to 100% for elution of hydrophobic cations and column cleaning
  • Use conductivity or UV detection
Product Family Options
Product Information
  • Phase
    Column Format
    Column hardware
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • PRP-X200
    Stainless steel
    10 µm
    2,10 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 956.00
    SFr. 956.00
    1 * 1 ST
       SFr. 956.00
  • PRP-X200
    Stainless steel
    10 µm
    4,10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 956.00
    SFr. 956.00
    1 * 1 ST
       SFr. 956.00
  • PRP-X200
    Stainless steel
    10 µm
    4,10 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’030.00
    SFr. 1’030.00
    1 * 1 ST
       SFr. 1’030.00
  • PRP-X200
    Stainless steel
    10 µm
    4,10 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’090.00
    SFr. 1’090.00
    1 * 1 ST
       SFr. 1’090.00
  • PRP-X200
    PEEK
    10 µm
    4,60 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’080.00
    SFr. 1’080.00
    1 * 1 ST
       SFr. 1’080.00
  • PRP-X200
    PEEK
    10 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 1’200.00
    SFr. 1’200.00
    1 * 1 ST
       SFr. 1’200.00
Analytical guard columns
Product Information
  • Phase
    Column Format
    Column hardware
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • 554-1270
    Analytical guard column starter kit (1 holder, 2 cartridges)
    Analytical guard column starter kit (1 holder, 2 cartridges)
    PRP-X200
    Guard column
    Stainless steel
    10 µm
    2,00 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 384.00
    SFr. 384.00
    1 * 1 KIT
       SFr. 384.00
  • 554-1264
    Analytical guard column replacement cartridges
    Analytical guard column replacement cartridges
    PRP-X200
    Guard column
    Stainless steel
    10 µm
    2,00 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 424.00
    SFr. 424.00
    1 * 5 ST
       SFr. 424.00
  • 554-1245
    Analytical guard column starter kit (1 holder, 2 cartridges)
    Analytical guard column starter kit (1 holder, 2 cartridges)
    PRP-X200
    Guard column
    PEEK
    10 µm
    3,00 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 467.00
    SFr. 467.00
    1 * 1 ST
       SFr. 467.00
  • 554-1246
    Analytical guard column replacement cartridges
    Analytical guard column replacement cartridges
    PRP-X200
    Guard column
    PEEK
    10 µm
    3,00 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 519.00
    SFr. 519.00
    1 * 5 ST
       SFr. 519.00
  • HAMI79288
    Analytical guard column
    Analytical guard column
    PRP-X200
    Guard column
    Stainless steel
    10 µm
    2,10 mm
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 534.00
    SFr. 534.00
    1 * 1 ST
       SFr. 534.00
Semi-Preparative/preparative guard columns
Product Information
  • Phase
    Column Format
    Column hardware
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • 554-1227
    Semipreparative/preparative guard column starter kit (1 holder, 1 cartridge)
    Semipreparative/preparative guard column starter kit (1 holder, 1 cartridge)
    PRP-X200
    Guard column
    Stainless steel
    12 - 20 µm
    4,60 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 600.00
    SFr. 600.00
    1 * 1 ST
       SFr. 600.00
  • 554-1228
    Semipreparative/preparative guard column replacement cartridges
    Semipreparative/preparative guard column replacement cartridges
    PRP-X200
    Guard column
    Stainless steel
    12 - 20 µm
    4,60 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - SFr. 657.00
    SFr. 657.00
    1 * 2 ST
       SFr. 657.00
  • HAMI79914
    Semipreparative/preparative guard column
    Semipreparative/preparative guard column
    PRP-X200
    Guard column
    Stainless steel
    12 - 20 µm
    4,60 mm
    33 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 534.00
    SFr. 534.00
    1 * 1 ST
       SFr. 534.00