VWR® Hichrom Cutters, Polymeric Tubing
Supplier: Avantor
Certificates
About this item
The Clean-cut™ cutter is designed specifically for cutting polymeric HPLC tubing from 1/16” to 1/8” with no distortion of cross-section and without closing the bore.
- Cuts PEEK and PTFE tubing
- Square, burr-free cuts
- Convenient, replaceable blade
The “Guillotine” cutter is an alternative design for cutting polymeric tubing of 1/16” to 1/2”. A razor-sharp blade cuts while a V-shaped grove on the handle holds the tubing at the right angle to produce a square cut.
Frequently Bought Together