Orders LinkContactUsLinkComponent
Clamp with chain
Supplier:   BOCHEM
Clamp with chain
Supplier:  BOCHEM
Certificates
Clamp with chain
Clamp with chain
2 Options Available Below
Product Details & Documents
For attaching glass beakers, flasks, reaction vessels and other laboratory vessels to support stands.
  • Adjustable chain with silicone coating, 70 cm
  • Without bosshead for fixing to rod
Product Family Options
Product Information
  • Material
    Spanning width
    Length
    For
    Availability
    Price
  • Stainless steel 18/10
    50 - 160 mm
    250 mm
    Beakers
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 128.00
    SFr. 128.00
    1 * 1 ST
       SFr. 128.00
  • Galvanised steel
    50 - 160 mm
    250 mm
    Beakers
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 54.40
    SFr. 54.40
    1 * 1 ST
       SFr. 54.40