Clamps, three pronged
Catalog # 241-3156
Supplier: BOCHEM
Specifications
- Spanning width:0 - 70 mm
- Environmentally Preferable:
- Pk:1
Specifications
About this item
Aluminium.
- The fingers of the clamps are covered with a plastic coating to protect the items clamped
- Without bosshead for fixing to stand
- With T-screws screws for rapid easy fixing