ContactUsLinkComponent
VWR® Clamps, Three Pronged
VWR® Clamps, Three Pronged
Catalog # 241-0019
Supplier:  VWR Collection
undefined
VWR® Clamps, Three Pronged
Catalog # 241-0019
Supplier:  VWR Collection
Specifications
  • Spanning width:
    12 - 100 mm
  • Pk:
    5
Specifications
About this item
Laboral aluminium alloy.
  • Two easy action knurled screws for adjusting the clamping fingers independently of each other