ContactUsLinkComponent
VWR®, Clamps, Three Pronged, with Flexible Rod
VWR®, Clamps, Three Pronged, with Flexible Rod
Catalog # 241-0034
Supplier:  VWR Collection
undefined
VWR®, Clamps, Three Pronged, with Flexible Rod
Catalog # 241-0034
Supplier:  VWR Collection
Specifications
  • Spanning width:
    0 - 30 mm
  • Environmentally Preferable:
  • Pk:
    2
Specifications
About this item
Laboral aluminium alloy.
  • Jaws covered in vinyl to protect clamped items
  • Clamp rod diameter: 8 mm; rod length: 200 mm