Order Entry
Switzerland
ContactUsLinkComponent
VWR® Stand Clamps with Flexible Rod
VWR® Stand Clamps with Flexible Rod
Catalog # 241-0040
Supplier:  VWR Collection
undefined
VWR® Stand Clamps with Flexible Rod
Catalog # 241-0040
Supplier:  VWR Collection

Specifications

  • Spanning width:
    15 - 65 mm
  • Pk:
    2

Specifications

About this item

Laboral aluminium alloy. Clamp with flexible arm, mounting rod 12 mm Ø and rod length is 200 mm.

  • Round jaws covered in vinyl to protect clamped items
  • Convenient flexible arm has a maximum load of 500 g