Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bacterial cell spreaders

Bacterial cell spreaders

Bacterial cell spreader, Green, 180 mm, Individually packed
Catalog # 391-3015
Supplier:   Simport Scientific
Bacterial cell spreaders
Catalog # 391-3015
Supplier:  Simport Scientific
Bacterial cell spreaders
Bacterial cell spreaders

Bacterial cell spreader, Green, 180 mm, Individually packed

Price
SFr. 131.00
Log in for availability
1 * 100 ST
Product Details & Documents

PS, sterile.

  • No flame sterilisation needed
  • Designed for easy spreading of cells onto the surface of an agar plate
  • Supplied sterile in individual packs

Handle length: 176 mm
Head spreader width: 36 mm

Specifications
  • Packed:
    Individually packed
  • Colour:
    Green
  • Description:
    Bacterial cell spreader
  • Length:
    180 mm