Orders LinkContactUsLinkComponent
Cryobox, System 100™
Cryobox, System 100™
10×10, System 100™ cryotubes, 133×133×52 mm
Catalog # 479-3314
   Low Manufacturing Impact
Cryobox, System 100™
Catalog # 479-3314
   Low Manufacturing Impact
Cryobox, System 100™
Cryobox, System 100™
10×10, System 100™ cryotubes, 133×133×52 mm
Price
SFr. 12.00
Sale
Valid until 30/06/2026
Log in for availability
1 * 1 ST
Product Details & Documents
PC, autoclavable.
Designed for ultra-low temperature storage of  System 100™ cryotubes or other similar size, externally or internally threaded vials. For temperatures of –196 to +121 °C.
Transparent lid has printed grid and is keyed to prevent misalignment.
Specifications
  • Compartments:
    10×10
  • For:
    System 100™ cryotubes
  • W×D×H ext.:
    133×133×52 mm
  • Cat. No.:
    479-3314
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact