Bottles, wide neck, with screw cap, Nalgene®
Anbieter: Thermo Fisher Scientific
Zertifikate
Über diesen Artikel
FEP, translucent, with ETFE screw cap.
- Maximum chemical resistance
- Temperature resistant from −105 to +150 °C, leakproof
- Autoclavable
These bottles have the highest chemical and corrosion resistance. Suitable for applications at low and high temperatures, trace analyses, and for many types of work with organic solvents.
Technische Daten
- Catalog No:
- 215-7104
- 215-7101
- 215-7102
- 215-7103
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- ISO 14001 (Certificate Registration No. EMS 623400)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Technische Daten
Zugehörige Informationen
Frequently Bought Together