Bottles, wide neck, with screw cap
Supplier: KARTELL
Certificates
About this item
LDPE, translucent with LDPE screw cap.
- Good chemical resistance
- Heat resistant up to 75 °C
- With graduations, leakproof
Multipurpose bottles for a wide variety of uses. Easy filling and dispensing of powders, pastes and liquids.
DIN 13316 and DIN 168
Certifications: DIN 13316 and DIN 168
Frequently Bought Together