VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
Certificates
About this item
LDPE.
- Amber coloured to protect light-sensitive substances
- Leakproof, tough and versatile
Sample collection, storage and transport, for liquids, pastes and granular material. The wide mouth allows easy filling and cleaning.
Related Information
CoC VWR Bottles, Wide Neck, with Screw Cap.pdf
Conformity declaration
TDS_VWR_LDPE BOTTLES_AMBER_WIDE_MOUTH_WITH_SCREW_CAP.pdf
Product specification
VWR® Lab bric-a-brac
Product Literature
Frequently Bought Together