ContactUsLinkComponent
Wash bottles, wide neck, for microbiological staining solutions, Azlon®
Wash bottles, wide neck, for microbiological staining solutions, Azlon®
Catalog # 215-0595
Supplier:  SciLabware
undefined
Wash bottles, wide neck, for microbiological staining solutions, Azlon®
Catalog # 215-0595
Supplier:  SciLabware
Specifications
  • Description:
    Set of 5 wash bottles
  • Pk:
    5
Specifications
About this item
LDPE, with PP spray cap.
  • Set of five 250 ml venting wash bottles with DripLok® anti-drip valve
  • Printed for Lugols Iodine, Crystal Violet, Gram Colour Decolourising, Gram Iodine and Safranine Solutions
  • Printed GHS compliant hazard information where relevant and instructions
Certifications: Azlon® wash bottles meet the material requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.