Wash bottles, narrow neck, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
LDPE, translucent with PP spray cap.
- Good chemical resistance
- Heat resistant up to +80 °C
- Closure and stem moulded in one piece for leakproof performance
Economical, soft and very easy to press. For use with general solutions such as water, soap, dilute acids and dilute alkalis. The jet tip can be cut to increase the flow rate.
Specifications
- Catalog No:
- 215-8106
- 215-8105
- 215-8104
- 215-8103
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Specifications
Replacement caps
Product Information
Frequently Bought Together