Orders LinkContactUsLinkComponent
Screw Caps
Screw Caps
PP, black.
  • Very good chemical resistance
  • Heat-resistant up to 110 °C
  • Only suitable for bottles 215-2814 to 215-2819
Product Family Options
  • Thread
  • GL 18
    1 * 10 ST
    1 * 10 ST - SFr. 2.90
    SFr. 2.90
    1 * 10 ST
       SFr. 2.90
  • GL 25
    1 * 10 ST
    1 * 10 ST - SFr. 4.90
    SFr. 4.90
    1 * 10 ST
       SFr. 4.90
  • GL 28
    1 * 100 ST
    1 * 100 ST - SFr. 43.20
    SFr. 43.20
    1 * 100 ST
       SFr. 43.20
  • GL 32
    1 * 1 ST
    1 * 1 ST - SFr. 0.70
    SFr. 0.70
    1 * 1 ST
       SFr. 0.70
  • GL 40
    1 * 1 ST
    1 * 1 ST - SFr. 0.70
    SFr. 0.70
    1 * 1 ST
       SFr. 0.70
  • GL 45
    1 * 1 ST
    1 * 1 ST
    New Product
    New Product
  • GL 55
    1 * 100 ST
    1 * 100 ST - SFr. 78.20
    SFr. 78.20
    1 * 100 ST
       SFr. 78.20