Bottles, narrow neck, round, HDPE, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
HDPE, fluorinated with PP, fluorinated screw cap.
- Fluorinated bottles and carboys can be used for most acids, alkalis and aggressive organic solvents
- Heavy-duty walls are safe, durable and resistant to splitting and punctures
- Fluorocarbon surfaces inside and out
- Leakproof
Fluorinating these HDPE bottles and carboys provides a cost effective alternative to costly fluoropolymer containers.
Specifications
- Catalog No:
- 216-8502
- 216-8506
- 216-8501
- 216-8505
- 216-8504
- 216-8507
- 216-8503
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement