ContactUsLinkComponent
Replacement spigot for carboys and aspirator bottles
Replacement spigot for carboys and aspirator bottles
Catalog # 227-4009
Supplier:  KARTELL
undefined
Replacement spigot for carboys and aspirator bottles
Catalog # 227-4009
Supplier:  KARTELL
Specifications
  • Description:
    Replacement stopcock for carboys/aspirator bottles
  • Pk:
    1
Specifications
About this item
HDPE, white.
  • Good chemical resistance
  • Leakproof
  • Suitable for carboys/aspirator bottles 216-5744 to 216-5747
With ¾" BSP screw thread.