Carboys, wide neck, PP, vacuum, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PP, translucent with white PP screw cap (83B), TPE seal.
- Very good chemical resistance, heat resistant up to 135 °C
- Thick walled, heavy construction, rigid, robust, leakproof
- Maintains a total vacuum for more than 8 hours at 20 °C in vacuum applications
Ideal for use as vacuum bottles. For vacuum applications, please use the quick filling and venting screw caps. Cat. Nos. 216-4385 to 216-4388. The bottles are not designed for use with overpressure. They are well suited as transfer and filtering bottles as well as for biotechnical applications.
Specifications
- Catalog No:
- 216-8731
- 216-8732
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement