Carboys, narrow neck, PPCO, with screw cap, Nalgene®
PPCO, translucent with white PP screw cap.
- Very good chemical resistance
- Temperature resistant from −20 to +121 °C, autoclavable
- Leakproof
Large handle makes carrying easier while wearing rubber gloves for safe handling of corrosive liquids. Opening offset to one side for easy, precise pouring.
- Catalog No:
- 216-8681
- 216-8682
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together