VWR® Bossheads, Universal, Pivoting
Catalog # 241-0113
Anbieter: VWR Collection
Technische Daten
- Spanning width:3 - 16 mm
- Material:Laboral aluminium alloy
- Pk:2
Technische Daten
Über diesen Artikel
Bosshead made of laboral aluminium alloy for connecting two retort rods at any angle.
- Tommy screw for securing the bosshead at the desired angle
- T-screws for securing the retort rods