ContactUsLinkComponent
Measuring beakers with handle, Nalgene®
Measuring beakers with handle, Nalgene®
Catalog # 213-7154
   Sustainable Options Available
Supplier #: 1220-2500
Low Manufacturing Impact
undefined
Measuring beakers with handle, Nalgene®
Catalog # 213-7154
   Sustainable Options Available
Supplier #: 1220-2500
  Low Manufacturing Impact
Log in for availability
Select an Option
200 - 3000 ml
Select an Option
3000 ml
Select an Option
02
Select an Option
1
Specifications
  • Catalog No:
  • 213-7154
  • 213-7162
  • 213-7152
  • 213-7164
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
About this item
HDPE, translucent or PMP (TPX), crystal clear.
  • Good chemical resistance (HDPE)/Very good chemical resistance (PMP)
  • Good thermal resistance (HDPE)/Heat resistant up to +150 °C (briefly to +175 °C) (PMP)
  • Designed for rough handling (HDPE)/Autoclavable (PMP)
Dual graduations in ml and oz. Square handle for easy carrying and pouring. Extra strong wall for safer handling of acids, bases and other solutions. Transparent to microwaves (PMP).
Frequently Bought Together