ContactUsLinkComponent
Jacketed beakers, tall form
Jacketed beakers, tall form
Catalog # 213-0052
Supplier:  SciLabware
Supplier #: 1025/18SC
undefined
Jacketed beakers, tall form
Catalog # 213-0052
Supplier:  SciLabware
Supplier #: 1025/18SC
Log in for availability
Select an Option
95 mm
Select an Option
600 ml
Select an Option
180 mm
Select an Option
02
Select an Option
1
About this item
PYREX®, borosilicate glass, without spout.
  • Very good chemical resistance
  • Very good thermal resistance
  • With screw thread and plastic threaded tubing connectors
  • Tube nozzles with 11 mm ext. diameter (Ø) for 250 and 600 ml beakers and 14 mm ext. diameter (Ø) for 1000 and 2000 ml beakers
For maintaining liquid samples at a constant temperature. The temperature controlling medium is pumped through the double-walled jacket by means of a recirculating thermostat.
Frequently Bought Together