Orders LinkContactUsLinkComponent
Beakers, low form
Supplier:   NICKEL ELECTRO
Beakers, low form
Supplier:  NICKEL ELECTRO
Certificates
Beakers, low form
Beakers, low form
6 Options Available Below
Product Details & Documents
Aluminium, with spout.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 100 ml
    57 mm
    50,4 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 65.90
    SFr. 65.90
    1 * 5 ST
       SFr. 65.90
  • 250 ml
    76 mm
    69 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 74.50
    SFr. 74.50
    1 * 5 ST
       SFr. 74.50
  • 500 ml
    103 mm
    84,5 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - SFr. 33.30
    SFr. 33.30
    1 * 2 ST
       SFr. 33.30
  • 1000 ml
    132 mm
    102 mm
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - SFr. 42.00
    SFr. 42.00
    1 * 2 ST
       SFr. 42.00
  • 2000 ml
    156 mm
    126 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 40.40
    SFr. 40.40
    1 * 1 ST
       SFr. 40.40
  • 5000 ml
    206 mm
    177 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 66.70
    SFr. 66.70
    1 * 1 ST
       SFr. 66.70