ContactUsLinkComponent
Beakers, low form, Azlon®
  213-0453
 : BDD336P
undefined
Beakers, low form, Azlon®
  213-0453
 : BDD336P
Select an Option
87 mm
Select an Option
50 ml
Select an Option
500 ml
Select an Option
117 mm
Select an Option
10
Select an Option
6
PMP (TPX), crystal clear.
  • Excellent chemical resistance
  • Heat resistant up to +180 °C
  • Smooth surfaces for quick, easy cleaning
Ideal for general laboratory use. With spout and exceptionally easy to read red graduation. Straight sided.
ISO 7056
 : ISO 7056
Frequently Bought Together