Orders LinkContactUsLinkComponent
Beakers, low form
Beakers, low form
Supplier:   SciLabware
Beakers, low form
Supplier:  SciLabware
Certificates
Beakers, low form
Beakers, low form
15 Options Available Below
Product Details & Documents
PYREX® borosilicate glass, clear, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
Ideal for general laboratory use. With spout, white graduations and labelling area.
Certifications: ISO 3819 and DIN 12331 - Beaker 10000 ml supplementary to ISO range and graduations are approximate.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Availability
    Price
  • 5 ml
    30 mm
    22 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 93.30
    SFr. 93.30
    1 * 10 ST
       SFr. 93.30
  • 10 ml
    35 mm
    26 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 106.00
    SFr. 106.00
    1 * 10 ST
       SFr. 106.00
  • 25 ml
    50 mm
    34 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 90.80
    SFr. 90.80
    1 * 10 ST
       SFr. 90.80
  • 50 ml
    60 mm
    42 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 86.60
    SFr. 86.60
    1 * 10 ST
       SFr. 86.60
  • 100 ml
    70 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 86.20
    SFr. 86.20
    1 * 10 ST
       SFr. 86.20
  • 150 ml
    80 mm
    60 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 91.00
    SFr. 91.00
    1 * 10 ST
       SFr. 91.00
  • 250 ml
    95 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 93.60
    SFr. 93.60
    1 * 10 ST
       SFr. 93.60
  • 400 ml
    110 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 113.00
    SFr. 113.00
    1 * 10 ST
       SFr. 113.00
  • 600 ml
    125 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 140.00
    SFr. 140.00
    1 * 10 ST
       SFr. 140.00
  • 800 ml
    135 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 193.00
    SFr. 193.00
    1 * 10 ST
       SFr. 193.00
  • 1000 ml
    145 mm
    105 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 223.00
    SFr. 223.00
    1 * 10 ST
       SFr. 223.00
  • 2000 ml
    185 mm
    130 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 389.00
    SFr. 389.00
    1 * 10 ST
       SFr. 389.00
  • 3000 ml
    210 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 70.20
    SFr. 70.20
    1 * 1 ST
       SFr. 70.20
  • 5000 ml
    270 mm
    170 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 120.00
    SFr. 120.00
    1 * 1 ST
       SFr. 120.00
  • 10000 ml
    350 mm
    217 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 339.00
    SFr. 339.00
    1 * 1 ST
       SFr. 339.00