ContactUsLinkComponent
Beakers, low form, heavy duty
Beakers, low form, heavy duty
Catalog # 213-0048
Supplier:  SciLabware
Supplier #: 1003-1L
undefined
Beakers, low form, heavy duty
Catalog # 213-0048
Supplier:  SciLabware
Supplier #: 1003-1L
Log in for availability
Select an Option
108 mm
Select an Option
1000 ml
Select an Option
156 mm
Select an Option
05
Select an Option
6
About this item
Heavy duty beakers, PYREX® borosiicate glass. Mechanically stronger than standard beakers. Designed for longer life and increased user safety.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
Reinforced rim and spout. Less prone to breakage during washing etc. Thicker walls and heavier bases. White graduations and marking spot. With spout.
Frequently Bought Together