ContactUsLinkComponent
Beakers, low form, DURAN®
Beakers, low form, DURAN®
Catalog # 213-1105
Supplier:  DWK Life Sciences
Supplier #: 211062402
undefined
Beakers, low form, DURAN®
Catalog # 213-1105
Supplier:  DWK Life Sciences
Supplier #: 211062402
Log in for availability
Select an Option
50 mm
Select an Option
100 ml
Select an Option
70 mm
Select an Option
05
Select an Option
1
About this item
DURAN®, borosilicate glass 3.3, clear, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Graduated for approximate reading of contents and labelling area (213-1101, 213-1102 not graduated and without Retrace Code).
With printed Retrace Code (batch identification).
ISO 3819 (except 213-1115).
Frequently Bought Together