SP Bel-Art Biohazard Disposal Bags
Bright red polypropylene bags include black biohazard symbol and precautionary procedures printed in four languages (English, Spanish, French, German) for added safety.
Sterilisation indicator patch darkens when exposed to steam sterilisation for 20 minutes
- High temperature steam autoclavable to 135 °C (275 °F)
Type A: PP bags autoclavable to 135 °C, thickness: 38 µm.
Type B: Extra heavy duty bags blended, autoclavable to +121 °C, thickness: 50 µm.
Frequently Bought Together