Orders LinkContactUsLinkComponent
Adapters, delivery, with vent, QUICKFIT®
Supplier:   SciLabware
Adapters, delivery, with vent, QUICKFIT®
Supplier:  SciLabware
Certificates
Adapters, delivery, with vent, QUICKFIT®
Adapters, delivery, with vent, QUICKFIT®
2 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass, bend.
  • Avoids the problems of 'closed systems'
With a side arm for venting purposes, not for connection to a vacuum source. For adapters with vacuum connection, see 201-8653.
Product Family Options
Product Information
  • Socket size
    Cone size
    Availability
    Price
  • 14/23 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64.50 SFr.
    64.50 SFr.
    1 * 1 ST
       64.50 SFr.
  • 19/26 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62.50 SFr.
    62.50 SFr.
    1 * 1 ST
       62.50 SFr.