ContactUsLinkComponent
Adapters, receiver, vacuum, straight, Quickfit®
Adapters, receiver, vacuum, straight, Quickfit®
Catalog # 201-0605
Supplier:  SciLabware
undefined
Adapters, receiver, vacuum, straight, Quickfit®
Catalog # 201-0605
Supplier:  SciLabware
Specifications
  • Cone size:
    24/29 NS
  • Socket size:
    24/29 NS
  • Total length:
    140 mm
  • Pk:
    1
Specifications
About this item
PYREX®, borosilicate glass.
  • Delivery stem within cone
  • With side-arm for connection to a vacuum supply
  • Easy connection to flexible tubing via detachable plastic screwthread connector