ContactUsLinkComponent
Chambered cover glasses, Nunc™, Lab-Tek™
Chambered cover glasses, Nunc™, Lab-Tek™
Catalog # 732-3644
   Low Manufacturing Impact
Chambered cover glasses, Nunc™, Lab-Tek™
Catalog # 732-3644
   Low Manufacturing Impact
Specifications
  • Description:
    Coverglass for 16-well chamber slide
  • L×W:
    22×47 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    96
Specifications
About this item
These cover glasses provide secure content on a slide while viewing under a microscope.
For use with slide flask (NUNC170920) and Thermo Scientific™ Flaskette™ products.