ContactUsLinkComponent
IVF ICSI dish, Nunc™
IVF ICSI dish, Nunc™
Catalog # 392-0323
   Low Manufacturing Impact
undefined
IVF ICSI dish, Nunc™
Catalog # 392-0323
   Low Manufacturing Impact
Specifications
  • Ø:
    51 mm
  • Packed:
    Individually wrapped
  • Description:
    IVF ICSI dish, non treated, with lid
  • Height:
    10 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    120
Specifications
About this item
Non treated surface, sterile (SAL 10⁻⁶), with lid. This IVF ICSI dish meet the exacting requirements of IVF testing, certification, and validation, providing consistently reliable, high quality results during fertilisation.
  • Maintain optimal temperature with true flat bottom
  • Low profile provides optimal angles during ICSI process
  • Clearly see sample with super polished optical centre
  • Increased safety with easy grip design
  • Certificates of conformity available for every lot
Produced under ISO 13485 with full lot traceability
Certifications: Produced under ISO 13485 with full lot traceability.