ContactUsLinkComponent
Wash bottles, wide neck, colour coded, Unitary™, Nalgene®
Wash bottles, wide neck, colour coded, Unitary™, Nalgene®
Catalog # NALG2423-0500
   Low Manufacturing Impact
undefined
Wash bottles, wide neck, colour coded, Unitary™, Nalgene®
Catalog # NALG2423-0500
   Low Manufacturing Impact
Specifications
  • Thread:
    38-415
  • Capacity:
    500 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    4
Specifications
About this item
LDPE, translucent, with PP closure.
  • Good chemical resistance
  • Good temperature resistance
Coded Unitary LDPE wash bottles, which come in a convenient, pre-packaged and easy-to-identify assortment. Red, white, blue or yellow closures/stems on these flexible unitary bottles permit easy content identification.