Order Entry
Switzerland
ContactUsLinkComponent
Triethyl orthoacetate for synthesis, Sigma-Aldrich®
Flammable
Triethyl orthoacetate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Triethyl orthoacetate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    162.22 g/mol
  • Boiling Point:
    142 °C (1013 hPa)
  • Flash Point:
    32 °C
  • Refractive Index:
    1.3958 (20 °C, 589 nm)
  • Explosion Limit:
    0.9 - 7.0 %(V)
  • Density:
    0.89 g/cm3 (20 °C)
  • Ignition Point:
    455 °C

Specification Test Results

Related Information

Frequently Bought Together