
Light green SF yellowish
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5141-20-8
:
Formula:C₃₇H₃₄N₂Na₂O₉S₃
MDL Number:MFCD00012121
:792.86 g/mol
Merck Index:14,5485
Melting Pt:∼ 288 °C (dec.)
EINECS:225-906-5
:GHS08
Storage Temperature:Ambient
Related Information
Frequently Bought Together