Order Entry
Switzerland
ContactUsLinkComponent
1,4-Dioxane 0.1% in aqueous solution Reag. Ph. Eur. 1032001
1,4-Dioxane 0.1% in aqueous solution Reag. Ph. Eur. 1032001
Catalog # 87820.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
1,4-Dioxane 0.1% in aqueous solution Reag. Ph. Eur. 1032001
Catalog # 87820.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :88.11 g/mol
EINECS:204-661-8
UN:1165
ADR:3,II
CAS Number:123-91-1
Formula:C₄H₈O₂
MDL Number:MFCD00006571