Order Entry
Switzerland
ContactUsLinkComponent
VWR® PTFE-Tapes
VWR® PTFE-Tapes
Catalog # 332362S
Supplier:  VWR Chemicals
undefined
VWR® PTFE-Tapes
Catalog # 332362S
Supplier:  VWR Chemicals

Specifications

  • Width:
    12,0 mm
  • Description:
    Sealing tape
  • Length:
    12,0 m
  • Pk:
    1 Roll

Specifications

About this item

Colour: White.