Human Recombinant BAFF (Animal free) (from E. coli)
Anbieter: Shenandoah Biotechnology
New Product
Zertifikate
Über diesen Artikel
B cell-attracting chemokine 1 (BCA-1), also known as CXCL13, is expressed at high levels in lymphoid tissues, such as the spleen, lymph nodes, and Peyer's patches. BCA-1 activates signaling through the receptor Burkitt lymphoma receptor 1 (BLR1) to chemoattract B cells.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Technische Daten
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q9Y275
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:TALL|THANK|BLyS
- Purity:≥90%
- Molecular weight:18,5 kDa
- Sequence:MHHHHHHLVP RAVQGPEETV TQDCLQLIAD SETPTIQKGS YTFVPWLLSF KRGSALEEKE NKILVKETGY FFIYGQVLYT DKTYAMGHLI QRKKVHVFGD ELSLVTLFRC IQNMPETLPN NSCYSAGIAK LEEGDELQLA IPRENAQISL DGDVTFFGAL KLL
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Technische Daten
Frequently Bought Together