Order Entry
Switzerland
ContactUsLinkComponent
Electronic Bowie and Dick Test, Routine Control, Vacuum Test, EBI 16
Electronic Bowie and Dick Test, Routine Control, Vacuum Test, EBI 16
Supplier:  EBRO
Certificates
undefined
Electronic Bowie and Dick Test, Routine Control, Vacuum Test, EBI 16
Supplier:  EBRO

Specifications

  • Catalog No:
  • EBRO1340-6697
  • 620-2785
  • Model:
  • EBI 16
  • Sensor:
  • PT1000, class A
  • Protection class:
  • IP 68
  • Recording intervals:
  • 1 s
  • Weight:
  • 500 g
  • Data memory - No. of readings:
  • 3×6960
  • Batteries:
  • Lithium, 3 V user-replaceable

Specifications

Related Information

Software and readout station

Product Information

Frequently Bought Together