VWR®, Karl Fischer NewGen Solvent E-FI for volumetric two-component KF titration
Catalog # 88436.290
Supplier: VWR Chemicals
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Description:NewGen Solvent E-FI for volumetric two-component Karl Fischer titration
- Seq:131
Specifications
About this item
Medium for volumetric two-component Karl Fischer titration (ethanol-based), Free of imidazole (free of CMR substances)
- free of imidazole
- free of CMR substances
Specification:
Suitability (for KF-Titration): complying
Water content: 0.010 - 0.020% (v/v)