Millex® Syringe Filters, PVDF Membrane
Supplier: Merck
Certificates
About this item
Syringe filters with hydrophobic PVDF membrane. Used in vacuum line protection, transducer protection and gas filtration. Available as either 25 mm or pump-driven Millex®-FG₅₀ 50 mm diameter filters.
Filters are either non sterile or sterilised with ethylene oxide.
Specifications
- Catalog No:
- MILFSLGVS25US
- MILFSLGVS25LS
- MILFSLGVS25XS
- MILFSLGVS25PS
- Max. operating pressure:
- 4,1 bar
- 4,1 bar
- 4,1 bar
- 4,1 bar
- Max. temperature:
- 121 °C
- 121 °C
- 121 °C
- 121 °C