ContactUsLinkComponent
Millex® Syringe Filters, PTFE Membrane
Millex® Syringe Filters, PTFE Membrane
Supplier:  Merck
Certificates
undefined
Millex® Syringe Filters, PTFE Membrane
Supplier:  Merck
Specifications
  • Catalog No:
  • MILFSLLG013SL
  • MILFSLLG025SS
  • Max. operating pressure:
  • 6,9 bar
  • 10 bar
  • Max. temperature:
  • 45 °C
  • 45 °C