Multiscreen® 96 Well Plate, PTFE Membrane
MILFMDRPN0410
: Merck
- Recommended working volume:1,8 ml
- Sterile:-
- Colour:Clear
- Description:96-well PTFE plate
- Environmentally Preferable:
- Pk:10
- Well volume:1,9 ml
The MultiScreen® deep well solvinert filter plate is intended for a wide range of applications including total drug analysis, bead cleavage (solid-phase combinatorial chemistry and peptide synthesis), and sample preparation before high-performance liquid chromatography (HPLC)/ultraviolet (UV) and liquid chromatography-tandem mass spectrometry (LC-MS/MS). This filter plate is fully automation compatible.
- H: 40,7 mm
- L: 128 mm
- W: 85,5 mm
- Filtration area: 0,28 cm²
- Hydrophobic
- Ambient
MultiScreen® deep, 96 well solvinert has been used to produce adsorption equilibrium isotherms. It has also been used in protein binding experiments.