ContactUsLinkComponent
Chemicon® BacterialXpress™ Nucleic Acid Extraction Kit
Chemicon® BacterialXpress™ Nucleic Acid Extraction Kit
Catalog # MILF3096
Supplier:  Merck
CAS Number:  
undefined
Chemicon® BacterialXpress™ Nucleic Acid Extraction Kit
Catalog # MILF3096
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    50
  • Description:
    BacterialXpress™ Nucleic acid extraction kit
Specifications
About this item
BacterialXpress™ Nucleic acid extraction kit is intended for the purification of bacterial DNA from plasma, serum, cerebral spinal fluid, amniotic fluid, tissue, bone marrow, and cell culture.
The BacterialXpress™ kit provides a rapid method for the extraction and purification of amplification-competent nucleic acids; however, the kit does not separate DNA from RNA. The BacterialXpress™ nucleic acid extraction kit can be used for the extraction and purification of a broad range of bacterial DNA from a variety of sources. However, performance cannot be guaranteed for all bacteria from all sources.
Suitable for the purification of bacterial DNA from plasma, serum, cerebral spinal fluid, amniotic fluid, tissue, bone marrow, and cell culture.
Delivery: Supplied woth Bacterial DNA extraction reagent (11 ml).