Auftragserfassung
Switzerland
ContactUsLinkComponent
Millex® MCE MF-Millipore™ Membrane Syringe Filters, Sterile
Millex® MCE MF-Millipore™ Membrane Syringe Filters, Sterile
Supplier:  Merck
Certificates
undefined
Millex® MCE MF-Millipore™ Membrane Syringe Filters, Sterile
Supplier:  Merck

Specifications

  • Catalog No:
  • MILFSLGSR33SS
  • MILFSLGSR33SB
  • MILFSLAAR33SS
  • MILFSLAAR33SB
  • MILFSLHAR33SS
  • Abmessungen:
  • 33×27 mm (Ø×H)
  • 33×27 mm (Ø×H)
  • 33×27 mm (Ø×H)
  • 33×27 mm (Ø×H)
  • 33×27 mm (Ø×H)
  • Haltevolumen:
  • <0,1 ml
  • <0,1 ml
  • <0,1 ml
  • <0,1 ml
  • <0,1 ml
  • Max. operating pressure:
  • 10,34 bar
  • 10,34 bar
  • 10,34 bar
  • 10,34 bar
  • 10,34 bar
  • Max. temperature:
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C

Specifications

Related Information

Frequently Bought Together