Cellulose Fiber Membrane Filters
: Merck
Cellulose support pads are used to reinforce filters in monitors for contamination analysis. When saturated with growth medium, they can also be used for culturing microorganisms on membranes.
- Filter code: AP10
- Filter type: Depth filter
- Hydrophilic
Applications include: Air monitoring, environmental monitoring, aerosol contamination monitoring, protecting membrane filters during high pressure or fast flow conditions.
: Comes in a pack of 100.
- Catalog No:
- MILFAP30034P0
- MILFAP1003700
- MILFAP1001300
- MILFAP1004700
- Thickness:
- 0,7874 - 0,9906 mm
- 0,7874 - 0,9906 mm
- 0,7874 - 0,9906 mm
- 0,7874 - 0,9906 mm
- Material:
- Cellulose
- Cellulose
- Cellulose
- Cellulose
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
Frequently Bought Together