ContactUsLinkComponent
Millex® Syringe Filters, LCR Hydrophilic PTFE
Millex® Syringe Filters, LCR Hydrophilic PTFE
Supplier:  Merck
Certificates
undefined
Millex® Syringe Filters, LCR Hydrophilic PTFE
Supplier:  Merck
Specifications
  • Catalog No:
  • MILFSLCRX13TL
  • MILFSLCRX13NL
  • MILFSLCRX13NK
  • MILFSLCR033NB
  • MILFSLLGX13NL
  • MILFSLLGX13NK
  • MILFSLLG033NK
  • MILFSLLG033NB
  • MILFSLLG033NS
  • MILFSLCR033NS
  • MILFSLCR033NK
  • Hold-up volume:
  • ≤15 µl after air purge
  • ≤15 µl after air purge
  • ≤15 µl after air purge
  • ≤80 µl after air purge
  • ≤15 µl after air purge
  • ≤15 µl after air purge
  • ≤80 µl after air purge
  • ≤80 µl after air purge
  • ≤80 µl after air purge
  • ≤80 µl after air purge
  • ≤80 µl after air purge
  • Max. operating pressure:
  • 10 bar
  • 10 bar
  • 10 bar
  • 8,6 bar
  • 10 bar
  • 10 bar
  • 8,6 bar
  • 8,6 bar
  • 8,6 bar
  • 8,6 bar
  • 8,6 bar
  • Max. temperature:
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • 45 °C
  • Water flow rate:
Specifications